4-(Methylthio)benzylamin

Lieferant: Wako

Synonyme: (4-(Methylthio)phenyl)methanamine

358-14563
WAKO358-14563EA 635 EUR
WAKO358-14563
4-(Methylthio)benzylamin
4-(Methylthio)benzylamin
Formel: C₈H₁₁NS
Molecular Weight: 153,2500 g/mol
MDL: MFCD01310831
CAS-Nummer: 83171-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR