Peptide standard mixture, MassPREP™
Lieferant: WATERS
The MassPREP™ peptide standard mixture is useful for the evaluation of HPLC columns and systems dedicated for peptide separations.
Each standard contains a void volume marker and nine, carefully selected peptides with a broad range of polarities and isoelectric points. All peaks are easily resolved under reversed-phase HPLC conditions typically used for peptide separations.