SPE cartridge, Sep-Pak®
Lieferant: WATERS
WAT037500
WATEWAT037500EA 705 EUR
WATEWAT037500
SPE cartridge, Sep-Pak®
Chromatographiesäulen
Specially designed for analysing formaldehyde, other aldehydes and ketones in air.