Standard, erbium perchlorate solution, for UV wavelength accuracy check
Lieferant: WATERS
WAT042885
WATEWAT042885EA 230 EUR
WATEWAT042885
Standard, erbium perchlorate solution, for UV wavelength accuracy check
Standards Farbreferenzen und Spektroskopie-Standards
Lieferumfang: Kit contains standards appropriate for assessment of UV Detector. 1×10 ml standard solution contains: 10.0 gm/l erbuim perchlorate +/-5%) (purity >=99.9%) in 100% HPLC Grade Water