Wright-Giemsa stain modifiziert
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Wright-Giemsa Solution
M-Clarity™ Qualitätsstufe = MQ500
WG128-4L
WG128-4LEA
558
EUR
WG128-4L
Wright-Giemsa stain modifiziert
Wright-Giemsa stain (solutions & kits)
When blood films are stained using Wright-Giemsa Stain, the white blood cell nucleus and cytoplasm take on the characteristic blue or pink coloration. The combination of purified eosin and thiazine dyes in the product eliminates inconsistent staining and yields reproducible chromogenic responses.
Spezifikation Testergebnisse
Appearance (Turbidity) Essentially free of precipitate |
Clear |
Appearance (Colour) | Blue |
Appearance (Form) | Solution |
pH | 6.7 - 6.9 |
18 months |
Conforms Cells stain within the performance characteristics when using Sigma-Aldrich Procedure No. WG |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...