Grade 0903 filter papers for technical use, Whatman™
Lieferant: Whatman products (Cytiva)
10334885
WHAT10334885EA 526 EUR
WHAT10334885
Grade 0903 filter papers for technical use, Whatman™
Filter Filterpapiere Qualitatives Filterpapier
Thin filter paper with smooth surface. Offers medium to slow flow rate and good retention for small particles.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.