Grade 0905 filter papers for technical use, Whatman™
Lieferant: Whatman products (Cytiva)
10334987
WHAT10334987EA 635 EUR
WHAT10334987
Grade 0905 filter papers for technical use, Whatman™
Filter Filterpapiere Qualitatives Filterpapier
A creped paper for coarse particles, offers a very high filtration speed.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.