Thermometer adapter, Wheaton®
Lieferant: DWK Life Sciences
165996
WHEA165996EA 128 EUR
WHEA165996
Thermometer adapter, Wheaton®
Adapter Adapter, Normschliff mit Schlaucholiven
This adapter has an O-ring that holds a thermometer or gas inlet tube securely. A grease-free WHEATON Connection® on one end provides an inert connection with external-threaded glass components.
The adapter can also be used with gas inlet tubing and will accommodate any tube with an OD of 5 to 7 mm.