Arsine generator apparatus, modified, WHEATON®
Lieferant: DWK Life Sciences
180023 180024
WHEA180023EA 284 EUR
WHEA180023 WHEA180024
Arsine generator apparatus, modified, WHEATON®
Aufschlusssysteme Geräte zum Nachweis von Spurenelementen
Modified version for arsine analysis which eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes which conforms to ASTM / USP and EPA specifications.
The freestanding system has a standard taper 24/40 Clear-Seal™ joint. Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable