Rubber stoppers for lyophilisation, WHEATON®
Lieferant: DWK Life Sciences
W224100-282 224100-203 W224100-193 W224100-093 W224100-202
WHEAW224100-282EA 1390 EUR
WHEAW224100-282 WHEA224100-203 WHEAW224100-193 WHEAW224100-093 WHEAW224100-202
Rubber stoppers for lyophilisation, WHEATON®
Stopfen Gummistopfen
Ultra pure chlorobutyl-isoprene rubber stoppers for lyophilisation.
  • Very low level of moisture adsorption and absorption of the preservatives in aqueous solution
  • Reduced possibility of the legs sticking to each other
  • Hardness of the lyophilization stopper is 46 Shore A
Designed for use with vials and serum bottles.