Evaporative concentrators, Kuderna-Danish, WHEATON®
Lieferant: DWK Life Sciences
297883
WHEA297883EA 704 EUR
WHEA297883
Evaporative concentrators, Kuderna-Danish, WHEATON®
Verdampfer Zentrifugalverdampfer
This unit is designed for the concentration of trace amounts of analytes in an organic solvent prior to analysis.
  • Smooth clear-seal joints eliminate the need for grease, reduces sample contamination
  • Consists of a 270 mm three-ball Snyder column, a 500 ml flask, and a 10 ml receiving vessel
  • Made of WHEATON 33 low extractable borosilicate glass meets USP Type I and ASTM E 438 Type I, Class A requirements