Cyanide distillation kit with Clear-Seal® joints, WHEATON®
Lieferant: DWK Life Sciences
377160
WHEA377160EA 1140 EUR
WHEA377160
Cyanide distillation kit with Clear-Seal® joints, WHEATON®
Destillationsbauteile Destillations-Bausätze
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • The sample is reacted with H₂SO₄ and MgCl₂, which converts any cyanide to HCN
  • HCN distills over and is absorbed in a NaOH solution, which is then analyzed for CN
Ideal for removing interfering substances in quantitative analysis for cyanide.
Standards: Standard Test Method for Cyanides in Water,# ASTM D2036