KIMBLE® Black Phenolic, Screw Thread Caps
Lieferant: DWK Life Sciences
73809-15425
WHEA73809-15425EA 0 EUR
WHEA73809-15425
KIMBLE® Black Phenolic, Screw Thread Caps
Verschlüsse Flaschenverschlüsse
Black phenolic cap has a securely mounted LDPE cone-shaped liner which offers a two part seal. The closure forms a seal around the rim and the polyseal cone forms a seal against the inner diameter of the vial opening.
  • Designed for superior torque retention
  • Stress crack resistant
  • Excellent for sample storage and re-sealing
  • Identified in ASTM Specification E982, Class A requirements
  • Continuous knurling