Pipette tips, WHEATON®
Lieferant: DWK Life Sciences
851358
WHEA851358EA 175 EUR
WHEA851358
Pipette tips, WHEATON®
Pipettenspitzen
These high quality tips are made of high grade, virgin, metal-free PP.
  • Compatible with most macro pipettes
  • Precision injection molding care for air-tight fit
  • Free from trace metals for excellent accuracy and reproducibility