Dispensing cannula for 5, 10 and 20 ml Dosys™ syringes, SOCOREX® DOSYS™
Lieferant: DWK Life Sciences
851412
WHEA851412EA 112 EUR
WHEA851412
Dispensing cannula for 5, 10 and 20 ml Dosys™ syringes, SOCOREX® DOSYS™
Kanülen
Blunt end dispensing cannulas with Luer Lock. Ideal for use with 24 and 96 well microplates.
  • Stainless steel
  • Autoclavable
This unique dispensing system enables you to perform sample inoculations and cell feedings faster and more efficiently under sterile conditions.