Complete sampler, Wheaton® E-Z
Lieferant: DWK Life Sciences
885020
WHEA885020EA 604 EUR
WHEA885020
Complete sampler, Wheaton® E-Z
Probenehmer Probenehmer für Flüssigkeiten
The filling ports are located above the bottle to ensure that it is completely filled to reduce the possibility of air entrapment.
  • Head/shaft assembly and borosilicate glass bottle with lined cap
  • Constructed of chemically resistant PP
  • Bottle with a GPI 33-430 thread
This sampler is an efficient and time saving method of sampling from drums and tanks.