Bombe zur Probenahme, Wheaton®
Lieferant: DWK Life Sciences
885200
WHEA885200EA 3080 EUR
WHEA885200
Bombe zur Probenahme, Wheaton®
Probenehmer Probenehmer für Flüssigkeiten
An efficient and unique composite sampling device made of stainless steel.
  • Cylindrical chamber
  • Weighted plunger that seals the chamber
  • Cable attachments for suspending the apparatus and activating the sampling device
Suitable for collecting liquid from top, bottom or any intermediate level of storage tanks, tank cars, lagoons, ponds and wells.
Standards: EPA-Methoden zur Mischprobenahme 'Testmethoden zur Untersuchung von Abfällen 1.1.4.1'.