WHEATON® Container, Wide Mouth
Lieferant: DWK Life Sciences
W209916 W209912
WHEAW209916EA 202 EUR
WHEAW209916 WHEAW209912
WHEATON® Container, Wide Mouth
Container made of polystyrene with foam PE lined polypropylene screw cap.
  • Clear polystyrene offers excellent calrity of contents within
  • Ideal for the storage of solid samples and powders
  • Good resistance to inorganic chemicals