Ultrapure lyophilisation stopper, WHEATON®
Lieferant: DWK Life Sciences
W224100-407
WHEAW224100-407EA 795 EUR
WHEAW224100-407
Ultrapure lyophilisation stopper, WHEATON®
Stopfen Gummistopfen
This ultrapure lyophilization stopper is a very low moisture adsortion stopper. Manufactured from an ultra pure bromobutyl formulation with extremely high chemical purity and low gas permeability.
  • Very low level of adsortion of the preservatives present in aqueous parenteral solutions
  • Autoclavable
  • Free of latex