WHEATON® COMPLETEPACK Sterile Components, Seals Only
Lieferant: DWK Life Sciences
W988009-S W988010-S W988011-S W988012-S
WHEAW988009-SEA 1290 EUR
WHEAW988009-S WHEAW988010-S WHEAW988011-S WHEAW988012-S
WHEATON® COMPLETEPACK Sterile Components, Seals Only
Verschlüsse Chromatographieampullen-Verschlüsse
Aluminium centre hole seals with polypropylene flip off caps, sterilised.
  • Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Complete manufacturing and certificate of analysis traceability
  • Ready-to-Use, Just-in-Time, minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
USP <788>/EP 2.9.19 particulate matter in injections (vials and stoppers), USP <85>/EP 2.6.1, The Bacterial Endotoxins Test (vials and stoppers) and lot certified to SAL <10⁻⁶ (vials, stoppers, and seals), USP <660>/EP 3.2.1 glass containers for pharmaceutical use, and USP <381>/EP 3.2.9 elastomeric closures used in parenteral preparations.