Sedimentiergefäße, Imhoff, WHEATON®
Lieferant: DWK Life Sciences
W990800
WHEAW990800EA 348 EUR
WHEAW990800
Sedimentiergefäße, Imhoff, WHEATON®
Trichter Sedimentiergefäße
Made of SAN.
For Volumetric determining the amount of settleable solids.
Graduation: 0 to 1 ml/0,1 ml; 1 to 10 ml/0,5 ml; 10 to 40 ml/1 ml; 40 to 100 ml/2 ml.