NMR sample tubes, SP Wilmad-LabGlass
Lieferant: SP Industries
516-CC-5
WILM516-CC-5EA 99.5 EUR
WILM516-CC-5
NMR sample tubes, SP Wilmad-LabGlass
Röhrchen NMR-Röhrchen
Pyrex® glass.
  • 50 µm gap between the inner and outer tubes
  • Ideal for variable temperature studies
These double layered tubes provide protection to toxic samples. They can also be used with a reference standard which is insoluable in the sample or may cause a reaction.