NMR tubes, PRECISION, with screw cap, SP Wilmad-LabGlass
Lieferant: SP Industries
535-TR-7 513-7TRA-7 335-TR-7 507-TR-7 507-TR-8 528-TR-7 528-TR-8 535-TR-8
WILM535-TR-7EA 44.7 EUR
WILM535-TR-7 WILM513-7TRA-7 WILM335-TR-7 WILM507-TR-7 WILM507-TR-8 WILM528-TR-7 WILM528-TR-8 WILM535-TR-8
NMR tubes, PRECISION, with screw cap, SP Wilmad-LabGlass
Röhrchen NMR-Röhrchen
Borosilicate glass, Class A, ASTM Type 1.
These tubes are commonly used in sample degasification these tubes can maintain vacuum quality of >10⁻⁴
Lieferumfang: Tube comes complete with open top screw cap and septum.