Test tube racks, SP Wilmad-LabGlass
Lieferant: SP Industries
820-A
WILM820-AEA 88.4 EUR
WILM820-A
Test tube racks, SP Wilmad-LabGlass
Gestelle Reagenzglasgestelle
PP, natural.
These lightweight racks maximise benchtop storage capacity and is virtually inert. Ideal for use with NMR and EPR tubes.