Burette, straight, class A, PTFE stopcock, SP Wilmad-LabGlass
Lieferant: SP Industries
LG-4011T-102
WILMLG-4011T-102EA 724 EUR
WILMLG-4011T-102
Burette, straight, class A, PTFE stopcock, SP Wilmad-LabGlass
Büretten
Burette, made of borosilicate glass, with funnel shaped top to accommodate a rubber stopper and glass tubing to fill from a delivery tip via suction.
  • Precision type burette
  • With durable coloured scale
  • With PS 2 mm stopcock with PTFE plug