Large Nitrogen Dewar Flask for Bruker
Lieferant: SP Industries
WG-853-B-Q
WILMWG-853-B-QEA 2510 EUR
WILMWG-853-B-Q
Large Nitrogen Dewar Flask for Bruker
Dewars und Kryogefäße
These dewar Flasks provide a convenient alternative to VT systems for studies at 77 K, and are silvered around the reservoir. 150 ml flask lasts 30 minutes after a filling. The offset versions (WG-853) are designed for cavities that have limited space between the reservoir and the drive shaft for iris.