Power selector, WRT00117
Lieferant: WINKLER
WRT00117-XX010
WINKWRT00117-XX010EA 147 EUR
WINKWRT00117-XX010
Power selector, WRT00117
Regler Temperaturregler
Plug-in power selector made of polystyrene with switching mechanism.
  • Rotary switch with bimetal element
  • Signal light for controlling status indication