Screw caps for thread connection
Lieferant: WITEG LABORTECHNIK
0.176.032
WITG0.176.032EA 3.1 EUR
WITG0.176.032
Screw caps for thread connection
Verschlüsse Flaschenverschlüsse
PBT, red without seals.
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air
With central hole.