Gaskets for screw caps
Lieferant: witeg Labortechnik
0.180.012
WITG0.180.012EA 11.3 EUR
WITG0.180.012
Gaskets for screw caps
Versiegeln Dichtungen
Silicone with PTFE washers
  • Temperature resistant from −200 to +250 °C
  • Very good chemical resistance
  • Easy handling
For threaded connection caps with hole and GL thread.