Flasks, round bottom, with two necks
Lieferant: witeg Labortechnik
0.754.250
WITG0.754.250EA 38.9 EUR
WITG0.754.250
Flasks, round bottom, with two necks
Kolben Rundkolben
Borosilicate glass 3.3 two necks, parallel side neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one parallel side neck. With matt white labelling area.