Trichter mit Normschliffkern
Lieferant: witeg Labortechnik
1.650.014 1.650.029 1.650.129 1.650.130 1.650.150 1.650.019
WITG1.650.014EA 9.8 EUR
WITG1.650.014 WITG1.650.029 WITG1.650.129 WITG1.650.130 WITG1.650.150 WITG1.650.019
Trichter mit Normschliffkern
Trichter Filtertrichter Trichter mit Kernschliff
Funnels, made of boroslicate glass, with standard ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable