Trichter mit Normschliffkern
Lieferant: witeg Labortechnik
1.651.019 1.651.029
WITG1.651.019EA 66.1 EUR
WITG1.651.019 WITG1.651.029
Trichter mit Normschliffkern
Trichter Filtertrichter Trichter mit Kernschliff
Funnels, made of boroslicate glass, with standard ground joint cone, one side flat.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.