Dosing funnels, with spiral conveyor
Lieferant: WITEG LABORTECHNIK
1.897.825
WITG1.897.825EA 659 EUR
WITG1.897.825
Dosing funnels, with spiral conveyor
Trichter Tropftrichter
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For powdery substances. Horizontal version with spiral conveyor.