Scheidetrichter, rund
Lieferant: witeg Labortechnik
2.620.050
WITG2.620.050EA 108 EUR
WITG2.620.050
Scheidetrichter, rund
Trichter Scheidetrichter
Separatory funnel, globe (ball) shaped, made of borosilicate glass 3.3, with detachable PTFE-stopcock and PE stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance