Gas generators, Duran®
Lieferant: WITEG LABORTECHNIK
2.989.500
WITG2.989.500EA 374 EUR
WITG2.989.500
Gas generators, Duran®
Gasflaschen
Gas generators according to Kipp, Duran.
  • Tubulature at bottom
  • Jointless, total grease free apparatus
With two screw threads GL 25, one screw thread GL 45 and GL 18, with needle valve stopcock with overtwist device.
Lieferumfang: Gas generators complete with safety funnel, caps and seals, hole disc made of sterilisable silicone, 500 ml capacity of middle bulb.