Cuvette Washers
Lieferant: witeg Labortechnik
2.990.111
WITG2.990.111EA 0 EUR
WITG2.990.111
Cuvette Washers
Reinigungs- und Desinfektionsautomaten
Cuvette washers acc. to Swiegot, wash and dry cuvettes, cells, test tubes or small scale volumetric instruments with outer dimensions not larger than approx. 20 mm.
Activate flushing by lightly pressing inverted cell onto silicone gashet, complete with laboratory bottle GL 45, 500 ml, complete autoclavable.