Bottle-top dispensers, LABmax
Lieferant: witeg Labortechnik
5.370.001 5.370.801 5.370.802 5.370.803 5.370.804 5.370.805 5.370.002 5.370.003 5.370.004 5.370.005 5.370.006
613-9111EA 269 EUR
613-9111 WITG5.370.801 WITG5.370.802 WITG5.370.803 WITG5.370.804 WITG5.370.805 WITG5.370.002 WITG5.370.003 WITG5.370.004 WITG5.370.005 WITG5.370.006
Bottle-top dispensers, LABmax
Flaschenaufsatz-Dispenser
Dispensers have a unique direct displacement piston system that is chemical-resistant and provides safe, smooth dispensing.
  • Unique closed-circuit air purging system
  • Reagent guard valve prevents leakage
  • Exclusive 360° swivel turn
  • Quick, accurate volume adjustment
  • Unique piston design
Units are designed with a unique air purging system that prevents loss of reagents and eliminates dripping or splashing of media. An exclusive reagent guard valve prohibits accidental discharge and eliminates the need for capping the discharge tube when the dispenser isMore Product Information