Erlenmeyer flasks with screw caps
Lieferant: witeg Labortechnik
5.507.003 5.507.006 5.507.004 5.507.005 5.507.007
WITG5.507.003EA 18.1 EUR
WITG5.507.003 WITG5.507.006 WITG5.507.004 WITG5.507.005 WITG5.507.007
Erlenmeyer flasks with screw caps
Kolben Erlenmeyerkolben
Erlenmeyer flasks, made of borosilicate glass, with screw-thread, graduated can be used for the storage of culture media and mixing.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes