Flasks, flat bottom, narrow neck, with beaded rim
Lieferant: witeg Labortechnik
5.514.010
WITG5.514.010EA 269 EUR
WITG5.514.010
Flasks, flat bottom, narrow neck, with beaded rim
Kolben Stehkolben
DURAN® borosilicate glass 3.3 clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area.