Evaporating dishes with spout
Lieferant: witeg Labortechnik
5.530.010.B
WITG5.530.010.BEA 9.4 EUR
WITG5.530.010.B
Evaporating dishes with spout
Schalen/Schüsseln/Töpfe Abdampfschalen
Evaporating dishes are made of borosilicate glass, with spout.
  • Excellent chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standards: DIN 12336.