Vacuum desiccators, complete
Lieferant: WITEG LABORTECHNIK
5.661.250
WITG5.661.250EA 894 EUR
WITG5.661.250
Vacuum desiccators, complete
Exsikkatoren Exsikkatoren, Glas
Borosilicate glass 3.3. This vacuum desiccator has 10.5 litre capacity.
Lieferumfang: Supplied with two manometers, lid and side tube ST 24/29, PTFE needle valve stopcocks and with ring nut which has a safety coating.