Pulvertrichter
Lieferant: witeg Labortechnik
5.790.004 5.790.001
WITG5.790.004EA 18.2 EUR
WITG5.790.004 WITG5.790.001
Pulvertrichter
Trichter Pulvertrichter
Powder funnels, made of DURAN® borosilicate glass 3.3, with short wide stem.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bestellen
TECHNISCHE DATEN
Environmentally Preferable