Woulff bottles
Lieferant: witeg Labortechnik
5.801.001 5.801.500
WITG5.801.001EA 120 EUR
WITG5.801.001 WITG5.801.500
Woulff bottles
Flaschen Woulffsche Flaschen
Made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
With two side necks GL 18, center neck GL 25, complete with screw caps.