Multi-position magnetic hotplate stirrers
Lieferant: WITEG LABORTECHNIK
DH.WMH03506
WITGDH.WMH03506EA 2190 EUR
WITGDH.WMH03506
Multi-position magnetic hotplate stirrers
Rührer Mehrstellen-Magnetrührer mit Heizung
Digital multi-position magnetic stirrer ideal for mixing and heating simultaneously. Ideal for heat sensitive procedures in microbiology, biochemistry or sample drying. Units feater a powder-coated steel body which is easy to clean. Each plate can accommodate a support rod.
  • Each plate is individually adjustable via digital touch-button Jog-Dial controller
  • Heat and chemical resistant and resistant to certain acids
  • Timer: 99 h 59 min. with signal at end
  • Up to three temperature probes can be connected
Lieferumfang: Supplied with 30 mm magnetic stirring bar.