Rockers, digital, progammable, WiseMix®, RK
Lieferant: witeg Labortechnik
DH.WRK01010 DH.WRK01020
WITGDH.WRK01010EA 467 EUR
WITGDH.WRK01010 WITGDH.WRK01020
Rockers, digital, progammable, WiseMix®, RK
Schüttler Rotations-/Wippschüttler
Digital rocking shaker with programmable digital feedback control, smooth start, change of stirring speed, wave motion and high precision digital speed control.
  • Digital LCD display with back-light function for timer, RPM and power bar-graph
  • Over-load protection functions for motor
  • Includes 10 steps and 6 operational programs
  • Storage and alarm functions
  • Locking mode for experimental safety
  • Jog-Shuttle control system (patented) and CE certified
It can be used in gel staining, destaining, bblotting, cell culture, hybridisation and washing.