Polysorbat 20, J.T.Baker®

Lieferant: Avantor

X251-07EA 387.47 EUR
X251-07 X251-09
Polysorbat 20, J.T.Baker®
Polysorbat 20
Formel: C₅₈H₁₁₄O₂₆
Dichte: 1,1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00165986
CAS-Nummer: 9005-64-5
EINECS: 500-018-3

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use Passes Test
Identification (by IR) Passes Test
Not intended for Injectable Drug Use.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR