Amberlite® XAD-16, Porengröße: 200 Å 20 - 60 mesh Ionenaustauscherharz, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ400
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
- Surface area: 800 m²/g
Applications: Hydrophobic compounds up to 40,000 MW; antibiotics; pharmaceutical manufacturing; surfactants; bitters; separation of large organic molecules (especially proteins). More efficient than XAD2.
Formel:
C₁₀H₁₀ |
MDL:
MFCD00145831 CAS-Nummer: 104219-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...