Conductivity cells, 3200 series
Lieferant: YSI ENVIRONMENTAL
003252
YSIA003252EA 764 EUR
YSIA003252
Conductivity cells, 3200 series
Elektroden Leitfähigkeitselektroden
YSI 3200 series conductivity cells have built-in thermistors, allowing automatic temperature compensation. All YSI cells are calibrated according to the International Organization of Legal Metrology recommendations 56 and 68.
  • Made of ABS plastic or glass
  • For use with YSI 3200 series conductivity meters
  • Shipped with greater than 1% cell accuracy; includes certificate of traceability
  • Can be used as secondary lab standard
  • Cells can be re-calibrated (adjusted) to NIST traceable standards