Confidence solution, 5580
Lieferant: YSI ENVIRONMENTAL
005580
YSIA005580EA 222 EUR
YSIA005580
Confidence solution, 5580
Elektrolyte Elektrodenfülllösungen
This Confidence Solution® helps users to ensure probe performance of their pH, ORP, or conductivity probes.
This solution is designed for verifying measurements, not calibration. Simply immerse your multi-parameter probes in the solution and verify the pH, ORP, and conductivity values instantly. Ideal for quality checks during or after your study.
Expiration dates: Unopened=1 year, opened=3 months.
Lieferumfang: Case includes 6 plastic bottles (1 pint each).