Conductivity cells, 3400 series
Lieferant: YSI ENVIRONMENTAL
603401
YSIA603401EA 1360 EUR
YSIA603401
Conductivity cells, 3400 series
Elektroden Leitfähigkeitselektroden
Dip type conductivity cells made from 7740 PYREX borosilicate glass. For use with the 3100 and 3200 conductivity benchtop instruments (cell adapter required).